Product Details

SNP ID
rs201244384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39606384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCAGCTCGCCTTCCTCCTTGAC[A/C]TCGGCCAACGTGGCCTCCGTCCCCT
Phenotype
MIM: 601725
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NEUROD2 PubMed Links

Gene Details

Gene
NEUROD2
Gene Name
neuronal differentiation 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006160.3 416 Missense Mutation GAG,GAT E72D NP_006151.3

View Full Product Details