Product Details

SNP ID
rs201519432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28906507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCTCTTCGTCCTGTTTCTGGTG[C/T]CGGCGGCGCCCTGGGAAAAAGGGGG
Phenotype
MIM: 616157 MIM: 131560
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHRS13 PubMed Links

Gene Details

Gene
DHRS13
Gene Name
dehydrogenase/reductase 13
There are no transcripts associated with this gene.

Gene
FLOT2
Gene Name
flotillin 2
There are no transcripts associated with this gene.

Gene
PHF12
Gene Name
PHD finger protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033561.1 2530 Silent Mutation CGA,CGG R897R NP_001028733.1
NM_001290131.1 2530 Intron NP_001277060.1
NM_020889.2 2530 Intron NP_065940.1
XM_005258015.3 2530 Silent Mutation CGA,CGG R807R XP_005258072.1
XM_005258016.3 2530 Silent Mutation CGA,CGG R680R XP_005258073.1
XM_011525078.2 2530 Silent Mutation CGA,CGG R875R XP_011523380.1
XM_011525079.2 2530 Silent Mutation CGA,CGG R864R XP_011523381.1
XM_011525082.2 2530 Intron XP_011523384.1
XM_011525083.2 2530 Intron XP_011523385.1
XM_017024903.1 2530 Silent Mutation CGA,CGG R915R XP_016880392.1
XM_017024904.1 2530 Silent Mutation CGA,CGG R866R XP_016880393.1

View Full Product Details