Product Details

SNP ID
rs201655951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80135499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGATCTGCTGCTTCAGATAAG[A/G]TCAGCAACTGAAAGTGAAGAAAGAA
Phenotype
MIM: 608546
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4A3 PubMed Links

Gene Details

Gene
EIF4A3
Gene Name
eukaryotic translation initiation factor 4A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014740.3 1342 Silent Mutation GAC,GAT D409D NP_055555.1
XM_011525522.1 1342 Silent Mutation GAC,GAT D388D XP_011523824.1

View Full Product Details