Product Details

SNP ID
rs200698567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82362189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCATGCGGTATGAGGAAGAGGG[A/C]ATGTCCTACCTCTACGCCTCCTGGA
Phenotype
MIM: 615647
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TEX19 PubMed Links

Gene Details

Gene
TEX19
Gene Name
testis expressed 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207459.3 436 Silent Mutation GGA,GGC G13G NP_997342.1
XM_017024638.1 436 Silent Mutation GGA,GGC G13G XP_016880127.1
XM_017024639.1 436 Silent Mutation GGA,GGC G13G XP_016880128.1

View Full Product Details