Product Details

SNP ID
rs200800513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4671857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTCCTGGGGAGGGGGCCCTGCC[A/G]CAGGGCTTTCCCCTTCCCTCTCCAC
Phenotype
MIM: 609455
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PELP1 PubMed Links

Gene Details

Gene
PELP1
Gene Name
proline, glutamate and leucine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278241.1 3360 Missense Mutation GCG,GTG A821V NP_001265170.1
NM_014389.2 3360 Missense Mutation GCG,GTG A1095V NP_055204.3

View Full Product Details