Product Details

SNP ID
rs200666462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2387114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGTGGCTCACTGCCACGGGCTG[C/T]GGGTACAAGGGCAGCTGGGAGCCCA
Phenotype
MIM: 603039 MIM: 611418
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MNT PubMed Links

Gene Details

Gene
MNT
Gene Name
MAX network transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020310.2 822 Silent Mutation CCA,CCG P512P NP_064706.1
XM_011523868.2 822 Silent Mutation CCA,CCG P520P XP_011522170.1
XM_011523869.1 822 Silent Mutation CCA,CCG P231P XP_011522171.1
XM_017024654.1 822 Silent Mutation CCA,CCG P231P XP_016880143.1
Gene
SGSM2
Gene Name
small G protein signaling modulator 2
There are no transcripts associated with this gene.

View Full Product Details