Product Details

SNP ID
rs200802612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31863468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGATCTGTCCACAGTTCTCAGGT[C/T]TACCAAGGGGGTGGTTCAATTCTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPRS PubMed Links

Gene Details

Gene
COPRS
Gene Name
coordinator of PRMT5 and differentiation stimulator
There are no transcripts associated with this gene.

Gene
UTP6
Gene Name
UTP6, small subunit processome component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018428.2 1800 Missense Mutation AAA,AGA K562R NP_060898.2
XM_005258012.2 1800 Missense Mutation AAA,AGA K441R XP_005258069.1
XM_011524997.2 1800 UTR 3 XP_011523299.1
XM_017024845.1 1800 UTR 3 XP_016880334.1

View Full Product Details