Product Details

SNP ID
rs201147676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76079459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTCTGGCCATCCCTCTTCAAC[A/G]TCAACTTGTCCAAGAAGGTTCAGGA
Phenotype
MIM: 608163 MIM: 603691 MIM: 604858 MIM: 610935
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOC7 PubMed Links

Gene Details

Gene
EXOC7
Gene Name
exocyst complex component 7
There are no transcripts associated with this gene.

Gene
GALR2
Gene Name
galanin receptor 2
There are no transcripts associated with this gene.

Gene
SRP68
Gene Name
signal recognition particle 68
There are no transcripts associated with this gene.

Gene
ZACN
Gene Name
zinc activated ion channel
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_180990.3 201 Missense Mutation ATC,GTC I40V NP_851321.2

View Full Product Details