Product Details

SNP ID
rs201183215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41854076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAATGGCTGCTCCTTCGATGCT[C/T]AGGACTTCTGGAGGCAAGCTTTCAA
Phenotype
MIM: 608778
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL10 PubMed Links

Gene Details

Gene
KLHL10
Gene Name
kelch like family member 10
There are no transcripts associated with this gene.

Gene
KLHL11
Gene Name
kelch like family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018143.2 1852 Silent Mutation CTA,CTG L597L NP_060613.1

View Full Product Details