Product Details

SNP ID
rs201796374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5106110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTTCTCTCCTGTATGAATTCTC[C/T]GATGCTGACCGAGGTTTGAGCTCTG
Phenotype
MIM: 194480 MIM: 616463
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928000 PubMed Links

Gene Details

Gene
LOC101928000
Gene Name
uncharacterized LOC101928000
There are no transcripts associated with this gene.

Gene
ZFP3
Gene Name
ZFP3 zinc finger protein
There are no transcripts associated with this gene.

Gene
ZNF232
Gene Name
zinc finger protein 232
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320952.1 1167 Missense Mutation CAG,CGG Q341R NP_001307881.1
NM_001320953.1 1167 Missense Mutation CAG,CGG Q323R NP_001307882.1
NM_001320954.1 1167 Missense Mutation CAG,CGG Q323R NP_001307883.1
NM_001320955.1 1167 Missense Mutation CAG,CGG Q322R NP_001307884.1
NM_014519.3 1167 Missense Mutation CAG,CGG Q350R NP_055334.2
XM_011524006.2 1167 Missense Mutation CAG,CGG Q302R XP_011522308.1
XM_017025021.1 1167 Missense Mutation CAG,CGG Q350R XP_016880510.1
XM_017025022.1 1167 Missense Mutation CAG,CGG Q274R XP_016880511.1
XM_017025023.1 1167 Missense Mutation CAG,CGG Q265R XP_016880512.1

View Full Product Details