Product Details
- SNP ID
-
rs201082358
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:39766042 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAGATGGGCGGGGGCTTGAGGAG[A/T]TCGTAAGAGCGGGGAACCTCCTTCA
- Phenotype
-
MIM: 606221
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IKZF3
PubMed Links
Gene Details
- Gene
- IKZF3
- Gene Name
- IKAROS family zinc finger 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001257408.1 |
1121 |
Missense Mutation |
GAA,GAT |
E392D |
NP_001244337.1 |
NM_001257409.1 |
1121 |
Missense Mutation |
GAA,GAT |
E353D |
NP_001244338.1 |
NM_001257410.1 |
1121 |
Missense Mutation |
GAA,GAT |
E339D |
NP_001244339.1 |
NM_001257411.1 |
1121 |
Missense Mutation |
GAA,GAT |
E283D |
NP_001244340.1 |
NM_001257412.1 |
1121 |
Missense Mutation |
GAA,GAT |
E244D |
NP_001244341.1 |
NM_001257413.1 |
1121 |
Missense Mutation |
GAA,GAT |
E205D |
NP_001244342.1 |
NM_001257414.1 |
1121 |
Missense Mutation |
GAA,GAT |
E292D |
NP_001244343.1 |
NM_001284514.1 |
1121 |
Missense Mutation |
GAA,GAT |
E179D |
NP_001271443.1 |
NM_001284515.1 |
1121 |
Missense Mutation |
GAA,GAT |
E179D |
NP_001271444.1 |
NM_001284516.1 |
1121 |
Missense Mutation |
GAA,GAT |
E179D |
NP_001271445.1 |
NM_012481.4 |
1121 |
Missense Mutation |
GAA,GAT |
E426D |
NP_036613.2 |
NM_183228.2 |
1121 |
Missense Mutation |
GAA,GAT |
E370D |
NP_899051.1 |
NM_183229.2 |
1121 |
Missense Mutation |
GAA,GAT |
E387D |
NP_899052.1 |
NM_183230.2 |
1121 |
Missense Mutation |
GAA,GAT |
E387D |
NP_899053.1 |
NM_183231.2 |
1121 |
Missense Mutation |
GAA,GAT |
E331D |
NP_899054.1 |
NM_183232.2 |
1121 |
Missense Mutation |
GAA,GAT |
E348D |
NP_899055.1 |
View Full Product Details