Product Details

SNP ID
rs201525670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4735171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGCGGTGAGGATGAAGATGAT[A/G]GCCAGGAGGCACAGGACTGGCACTG
Phenotype
MIM: 605398 MIM: 612383 MIM: 614312
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXCL16 PubMed Links

Gene Details

Gene
CXCL16
Gene Name
C-X-C motif chemokine ligand 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100812.1 1228 Silent Mutation GCC,GCT A232A NP_001094282.1
NM_022059.3 1228 Silent Mutation GCC,GCT A232A NP_071342.2
Gene
MED11
Gene Name
mediator complex subunit 11
There are no transcripts associated with this gene.

Gene
ZMYND15
Gene Name
zinc finger MYND-type containing 15
There are no transcripts associated with this gene.

View Full Product Details