Product Details

SNP ID
rs201049268
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36014379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAATGGAAGCTTTGACATCATGA[A/G]CTCTGTCTCTGCATCTGGAAGAAGC
Phenotype
MIM: 602494
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CCL23 PubMed Links

Gene Details

Gene
CCL23
Gene Name
C-C motif chemokine ligand 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005064.5 162 Missense Mutation CTC,TTC L31F NP_005055.3
NM_145898.3 162 Missense Mutation CTC,TTC L31F NP_665905.2

View Full Product Details