Product Details

SNP ID
rs201596402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10631615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGAAGGAGACGCCTTACGTCC[A/G]TGATGGCCTTCTTGGCCTTCTCCTC
Phenotype
MIM: 160720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYH3 PubMed Links

Gene Details

Gene
MYH3
Gene Name
myosin, heavy chain 3, skeletal muscle, embryonic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002470.3 6737 Missense Mutation NP_002461.2
XM_011523870.2 6737 Missense Mutation XP_011522172.1
XM_011523871.2 6737 Missense Mutation XP_011522173.1
Gene
MYHAS
Gene Name
myosin heavy chain gene cluster antisense RNA
There are no transcripts associated with this gene.

View Full Product Details