Product Details

SNP ID
rs201800581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41930725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCGTCTCTAAATCCGGTCACCA[C/T]GTCGGACCCCGAAGGCGAGACCTTG
Phenotype
MIM: 108728
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACLY PubMed Links

Gene Details

Gene
ACLY
Gene Name
ATP citrate lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001096.2 103 Intron NP_001087.2
NM_001303274.1 103 Intron NP_001290203.1
NM_001303275.1 103 Intron NP_001290204.1
NM_198830.1 103 Intron NP_942127.1
XM_005257395.1 103 Intron XP_005257452.1
XM_017024688.1 103 Intron XP_016880177.1
Gene
TTC25
Gene Name
tetratricopeptide repeat domain 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031421.3 103 Missense Mutation ACG,ATG T0M NP_113609.1
XM_011525323.2 103 UTR 5 XP_011523625.1
XM_011525324.2 103 Missense Mutation ACG,ATG T1M XP_011523626.1

View Full Product Details