Product Details
- SNP ID
-
rs201972895
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44320682 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACAGAAGTCTGGTCCTTCGGCCT[A/G]AACCCATTGAGCCAGCTCTTCACCA
- Phenotype
-
MIM: 605448
MIM: 610820
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RUNDC3A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2011951] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RUNDC3A
- Gene Name
- RUN domain containing 3A
There are no transcripts associated with this gene.
- Gene
- SLC25A39
- Gene Name
- solute carrier family 25 member 39
View Full Product Details