Product Details

SNP ID
rs200671413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41987881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAAGCTTCACTGCATTTGTGCA[C/G]TCTTCTATTGCATCATCTAGTTTCC
Phenotype
MIM: 601964
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC7 PubMed Links

Gene Details

Gene
DNAJC7
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144766.2 1119 Missense Mutation GAC,GAG D260E NP_001138238.1
NM_003315.3 1119 Missense Mutation GAC,GAG D316E NP_003306.3
XM_011525167.2 1119 Missense Mutation GAC,GAG D260E XP_011523469.1
XM_011525168.2 1119 Missense Mutation GAC,GAG D260E XP_011523470.1
XM_011525169.2 1119 Intron XP_011523471.1
XM_017024994.1 1119 Missense Mutation GAC,GAG D260E XP_016880483.1

View Full Product Details