Product Details

SNP ID
rs200842352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82079563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATGATCAGGTCCACGGCGGCTG[A/C]CACACGCTCCTCTAGGCCCTTCAGC
Phenotype
MIM: 600212
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FASN PubMed Links

Gene Details

Gene
FASN
Gene Name
fatty acid synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004104.4 7309 Missense Mutation GCA,TCA A2398S NP_004095.4
XM_011523538.2 7309 Missense Mutation GCA,TCA A2398S XP_011521840.1

View Full Product Details