Product Details

SNP ID
rs200842828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75261938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTGAAGGTTGCCCGAGGATGGT[C/T]GGGCCTGGCGTTGGGCGTGCGGCGG
Phenotype
MIM: 606006 MIM: 612072 MIM: 611974
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GGA3 PubMed Links

Gene Details

Gene
GGA3
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172703.2 271 Intron NP_001166174.1
NM_001172704.2 271 Intron NP_001166175.1
NM_001291641.1 271 Intron NP_001278570.1
NM_001291642.1 271 Intron NP_001278571.1
NM_014001.4 271 Intron NP_054720.1
NM_138619.3 271 Intron NP_619525.1
XM_011524563.2 271 Intron XP_011522865.1
XM_017024385.1 271 Intron XP_016879874.1
XM_017024386.1 271 Intron XP_016879875.1
XM_017024387.1 271 Intron XP_016879876.1
XM_017024388.1 271 Intron XP_016879877.1
XM_017024389.1 271 Intron XP_016879878.1
XM_017024390.1 271 Intron XP_016879879.1
XM_017024391.1 271 Intron XP_016879880.1
XM_017024392.1 271 Intron XP_016879881.1
Gene
LOC100287042
Gene Name
uncharacterized LOC100287042
There are no transcripts associated with this gene.

Gene
MIF4GD
Gene Name
MIF4G domain containing
There are no transcripts associated with this gene.

Gene
MRPS7
Gene Name
mitochondrial ribosomal protein S7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015971.3 271 Missense Mutation TCG,TTG S13L NP_057055.2

View Full Product Details