Product Details

SNP ID
rs200879332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42563214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTACTACCGTGGCGCTGTCGGT[G/T]GCACGTTTGACCGCCTGCACAACGC
Phenotype
MIM: 609855 MIM: 109684 MIM: 602976 MIM: 608665
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COASY PubMed Links

Gene Details

Gene
COASY
Gene Name
Coenzyme A synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042529.2 945 Missense Mutation GGC,TGC G198C NP_001035994.1
NM_001042532.3 945 Missense Mutation GGC,TGC G227C NP_001035997.2
NM_025233.6 945 Missense Mutation GGC,TGC G198C NP_079509.5
XM_006722116.3 945 Missense Mutation GGC,TGC G227C XP_006722179.1
XM_011525300.1 945 Missense Mutation GGC,TGC G198C XP_011523602.1
XM_017025167.1 945 Missense Mutation GGC,TGC G198C XP_016880656.1
XM_017025168.1 945 UTR 5 XP_016880657.1
Gene
HSD17B1
Gene Name
hydroxysteroid 17-beta dehydrogenase 1
There are no transcripts associated with this gene.

Gene
MLX
Gene Name
MLX, MAX dimerization protein
There are no transcripts associated with this gene.

Gene
PSMC3IP
Gene Name
PSMC3 interacting protein
There are no transcripts associated with this gene.

View Full Product Details