Product Details

SNP ID
rs200914420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63751976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTTTCTCAGGATCTTCCTCATT[C/T]ATGATTCGCTCCTTCTCTGCTCTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC47 PubMed Links

Gene Details

Gene
CCDC47
Gene Name
coiled-coil domain containing 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020198.2 1539 Missense Mutation ATA,ATG I445M NP_064583.2
XM_005257527.2 1539 Missense Mutation ATA,ATG I445M XP_005257584.1

View Full Product Details