Product Details

SNP ID
rs201255090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81715054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGCACTCTACAGCGGCCTGAG[C/T]GCCTCGCTGTGCAGACAGGTGCGTG
Phenotype
MIM: 602375 MIM: 606794
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL12 PubMed Links

Gene Details

Gene
MRPL12
Gene Name
mitochondrial ribosomal protein L12
There are no transcripts associated with this gene.

Gene
SLC25A10
Gene Name
solute carrier family 25 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270888.1 532 Silent Mutation AGC,AGT S65S NP_001257817.1
NM_001270953.1 532 Silent Mutation AGC,AGT S65S NP_001257882.1
NM_012140.4 532 Silent Mutation AGC,AGT S65S NP_036272.2
XM_017024220.1 532 Silent Mutation AGC,AGT S22S XP_016879709.1

View Full Product Details