Product Details

SNP ID
rs201256125
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79735066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCTGAGGGCTGGCTCCTTCTT[C/T]TACCCGGGCGGGAACGTCACCTACC
Phenotype
MIM: 616997
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ENPP7 PubMed Links

Gene Details

Gene
ENPP7
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178543.4 460 Silent Mutation TTC,TTT F141F NP_848638.3
XM_011524737.1 460 Silent Mutation TTC,TTT F214F XP_011523039.1

View Full Product Details