Product Details

SNP ID
rs200845614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:745455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTGCCACTCCTGAGGGCTGCAC[C/G]AAGGCATGACTTGCACCAGGGCCAC
Phenotype
MIM: 611627 MIM: 606969
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DBIL5P PubMed Links

Gene Details

Gene
DBIL5P
Gene Name
diazepam binding inhibitor-like 5, pseudogene
There are no transcripts associated with this gene.

Gene
FAM57A
Gene Name
family with sequence similarity 57 member A
There are no transcripts associated with this gene.

Gene
GEMIN4
Gene Name
gem nuclear organelle associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015721.2 2925 Missense Mutation TCG,TGG S863W NP_056536.2
XM_005256667.4 2925 Missense Mutation TCG,TGG S867W XP_005256724.1
XM_005256670.4 2925 Missense Mutation TCG,TGG S852W XP_005256727.1
XM_011523910.2 2925 Missense Mutation TCG,TGG S867W XP_011522212.1
XM_011523911.2 2925 Missense Mutation TCG,TGG S867W XP_011522213.1
XM_011523912.2 2925 Missense Mutation TCG,TGG S852W XP_011522214.1
XM_011523913.2 2925 Missense Mutation TCG,TGG S852W XP_011522215.1
XM_017024709.1 2925 Missense Mutation TCG,TGG S867W XP_016880198.1

View Full Product Details