Product Details

SNP ID
rs201874131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74749088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGGTGGTGAGCCGCATCCGCG[C/T]CGCACTCAACGCCGTGCGCCTGCTG
Phenotype
MIM: 609956 MIM: 604990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR3615 PubMed Links

Gene Details

Gene
MIR3615
Gene Name
microRNA 3615
There are no transcripts associated with this gene.

Gene
RAB37
Gene Name
RAB37, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
SLC9A3R1
Gene Name
SLC9A3 regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004252.4 477 Missense Mutation GCC,GTC A81V NP_004243.1

View Full Product Details