Product Details

SNP ID
rs202047426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35130349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCCATCCAGATCACCGTGCGGC[A/G]CAAGGAACCCCGGCAAAAGATCGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FNDC8 PubMed Links

Gene Details

Gene
FNDC8
Gene Name
fibronectin type III domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017559.2 972 Missense Mutation CAC,CGC H297R NP_060029.1
Gene
NLE1
Gene Name
notchless homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014445.2 972 Intron NP_001014445.1
NM_018096.4 972 Intron NP_060566.2
XM_017024777.1 972 UTR 3 XP_016880266.1
Gene
RAD51L3-RFFL
Gene Name
RAD51L3-RFFL readthrough
There are no transcripts associated with this gene.

View Full Product Details