Product Details

SNP ID
rs200645109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48964335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTCTTCTTCCCCTTTTGGGCCA[C/G]CAGCCAGTTCACAAAGTCTTGTTGG
Phenotype
MIM: 137240
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GIP PubMed Links

Gene Details

Gene
GIP
Gene Name
gastric inhibitory polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004123.2 330 Missense Mutation CTG,GTG L78V NP_004114.1

View Full Product Details