Product Details

SNP ID
rs201671292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30198486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATTCAAGCGGATGTCCCCACA[A/G]GGAATTTCTGTTGGTGTTTCTGGGG
Phenotype
MIM: 182138
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC6A4 PubMed Links

Gene Details

Gene
SLC6A4
Gene Name
solute carrier family 6 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001045.5 2439 Silent Mutation CCC,CCT P621P NP_001036.1

View Full Product Details