Product Details

SNP ID
rs201911699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40698540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAATATTAAAAAAAGTCTACCT[C/T]GTCCTTGACCAGAACAGCTTCCAGA
Phenotype
MIM: 607742
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT24 PubMed Links

Gene Details

Gene
KRT24
Gene Name
keratin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019016.2 1205 Missense Mutation CAA,CGA Q491R NP_061889.2
XM_006721739.3 1205 Missense Mutation CAA,CGA Q393R XP_006721802.1
XM_011524460.2 1205 Missense Mutation CAA,CGA Q388R XP_011522762.1
XM_017024299.1 1205 Intron XP_016879788.1
Gene
LOC105371775
Gene Name
uncharacterized LOC105371775
There are no transcripts associated with this gene.

View Full Product Details