Product Details

SNP ID
rs201946575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1646484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGCCCCCAGACAAAGGTGTTC[A/G]TGGAGGGCAGAACAGGGCGGATCAG
Phenotype
MIM: 607300 MIM: 607848 MIM: 607873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRPF8 PubMed Links

Gene Details

Gene
PRPF8
Gene Name
pre-mRNA processing factor 8
There are no transcripts associated with this gene.

Gene
RILP
Gene Name
Rab interacting lysosomal protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031430.2 1458 Silent Mutation CAC,CAT H388H NP_113618.2
XM_005256811.1 1458 Silent Mutation CAC,CAT H360H XP_005256868.1
XM_017025195.1 1458 UTR 3 XP_016880684.1
Gene
SCARF1
Gene Name
scavenger receptor class F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003693.3 1458 Intron NP_003684.2
NM_145350.2 1458 Intron NP_663325.1
XM_017025256.1 1458 Intron XP_016880745.1

View Full Product Details