Product Details
- SNP ID
-
rs202018008
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:48550522 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGAAAGGTGGTTGAGGCCATAGA[A/G]GGAGGGGCCGGCAGGGGGCGGCAGC
- Phenotype
-
MIM: 142967
MIM: 142966
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HOXB-AS1
PubMed Links
Gene Details
- Gene
- HOXB-AS1
- Gene Name
- HOXB cluster antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- HOXB2
- Gene Name
- homeobox B2
There are no transcripts associated with this gene.
- Gene
- HOXB3
- Gene Name
- homeobox B3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002146.4 |
11506 |
Missense Mutation |
CTC,TTC |
L370F |
NP_002137.4 |
XM_005257277.3 |
11506 |
Missense Mutation |
CTC,TTC |
L370F |
XP_005257334.1 |
XM_005257280.3 |
11506 |
Missense Mutation |
CTC,TTC |
L297F |
XP_005257337.1 |
XM_005257282.3 |
11506 |
Missense Mutation |
CTC,TTC |
L238F |
XP_005257339.1 |
XM_006721854.2 |
11506 |
Missense Mutation |
CTC,TTC |
L370F |
XP_006721917.1 |
XM_011524707.2 |
11506 |
Missense Mutation |
CTC,TTC |
L517F |
XP_011523009.1 |
XM_011524708.2 |
11506 |
Missense Mutation |
CTC,TTC |
L427F |
XP_011523010.1 |
XM_011524710.1 |
11506 |
Missense Mutation |
CTC,TTC |
L370F |
XP_011523012.1 |
XM_011524719.1 |
11506 |
Missense Mutation |
CTC,TTC |
L370F |
XP_011523021.1 |
XM_011524720.2 |
11506 |
Missense Mutation |
CTC,TTC |
L370F |
XP_011523022.1 |
XM_011524721.2 |
11506 |
Missense Mutation |
CTC,TTC |
L369F |
XP_011523023.1 |
XM_011524726.2 |
11506 |
Missense Mutation |
CTC,TTC |
L297F |
XP_011523028.1 |
XM_017024557.1 |
11506 |
Missense Mutation |
CTC,TTC |
L517F |
XP_016880046.1 |
XM_017024558.1 |
11506 |
Missense Mutation |
CTC,TTC |
L517F |
XP_016880047.1 |
XM_017024559.1 |
11506 |
Missense Mutation |
CTC,TTC |
L517F |
XP_016880048.1 |
XM_017024560.1 |
11506 |
Missense Mutation |
CTC,TTC |
L517F |
XP_016880049.1 |
XM_017024561.1 |
11506 |
Missense Mutation |
CTC,TTC |
L517F |
XP_016880050.1 |
XM_017024562.1 |
11506 |
Missense Mutation |
CTC,TTC |
L517F |
XP_016880051.1 |
XM_017024563.1 |
11506 |
Missense Mutation |
CTC,TTC |
L517F |
XP_016880052.1 |
View Full Product Details