Product Details

SNP ID
rs199889022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:13731702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATAGCAAGAAAGAGAAAAGAGT[G/T]TGAAGATGATCTTGTAAAGGAAAGT
Phenotype
MIM: 603514
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM210A PubMed Links

Gene Details

Gene
FAM210A
Gene Name
family with sequence similarity 210 member A
There are no transcripts associated with this gene.

Gene
RNMT
Gene Name
RNA guanine-7 methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308263.1 426 Missense Mutation TGT,TTT C62F NP_001295192.1
NM_003799.2 426 Missense Mutation TGT,TTT C62F NP_003790.1
XM_005258162.1 426 Missense Mutation TGT,TTT C62F XP_005258219.1
XM_011525749.1 426 Missense Mutation TGT,TTT C62F XP_011524051.1
XM_011525750.2 426 Missense Mutation TGT,TTT C62F XP_011524052.1
XM_011525751.1 426 Missense Mutation TGT,TTT C62F XP_011524053.1
XM_011525753.1 426 Intron XP_011524055.1
XM_017026061.1 426 Missense Mutation TGT,TTT C62F XP_016881550.1

View Full Product Details