Product Details
- SNP ID
-
rs200692446
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:13826236 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTACCACCACATCATGACGGCGAG[C/G]CGCTCAGGGGCCATCATCGCCGGCA
- Phenotype
-
MIM: 600042
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MC5R
PubMed Links
Gene Details
- Gene
- MC5R
- Gene Name
- melanocortin 5 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005913.2 |
693 |
Missense Mutation |
AGC,AGG |
S157R |
NP_005904.1 |
View Full Product Details