Product Details

SNP ID
rs199766619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:49490828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCAACTCCACCATTGTAACGTC[G/T]GAATGGTACACACTGTTTCTGTAAA
Phenotype
MIM: 603661
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C18orf32 PubMed Links

Gene Details

Gene
C18orf32
Gene Name
chromosome 18 open reading frame 32
There are no transcripts associated with this gene.

Gene
LOC101928144
Gene Name
uncharacterized LOC101928144
There are no transcripts associated with this gene.

Gene
MIR1539
Gene Name
microRNA 1539
There are no transcripts associated with this gene.

Gene
RPL17
Gene Name
ribosomal protein L17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000985.4 194 Silent Mutation AGA,CGA R61R NP_000976.1
NM_001035006.2 194 Intron NP_001030178.1
NM_001199340.1 194 Intron NP_001186269.1
NM_001199341.1 194 Intron NP_001186270.1
NM_001199342.1 194 Silent Mutation AGA,CGA R61R NP_001186271.1
NM_001199343.1 194 Silent Mutation AGA,CGA R61R NP_001186272.1
NM_001199344.1 194 Intron NP_001186273.1
NM_001199345.1 194 Intron NP_001186274.1
Gene
RPL17-C18orf32
Gene Name
RPL17-C18orf32 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199355.1 194 Silent Mutation AGA,CGA R61R NP_001186284.1
NM_001199356.1 194 Silent Mutation AGA,CGA R23R NP_001186285.1
Gene
SNORD58A
Gene Name
small nucleolar RNA, C/D box 58A
There are no transcripts associated with this gene.

Gene
SNORD58B
Gene Name
small nucleolar RNA, C/D box 58B
There are no transcripts associated with this gene.

Gene
SNORD58C
Gene Name
small nucleolar RNA, C/D box 58C
There are no transcripts associated with this gene.

View Full Product Details