Product Details
- SNP ID
-
rs200561867
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:50272885 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCAATTTGGAGGCAGAATCATCC[C/T]TGTTCTCCTCCTTGTCCTCCTCTGG
- Phenotype
-
MIM: 614759
MIM: 609150
MIM: 156535
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CFAP53
PubMed Links
Gene Details
- Gene
- CFAP53
- Gene Name
- cilia and flagella associated protein 53
There are no transcripts associated with this gene.
- Gene
- CXXC1
- Gene Name
- CXXC finger protein 1
There are no transcripts associated with this gene.
- Gene
- MBD1
- Gene Name
- methyl-CpG binding domain protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001204136.1 |
1885 |
Missense Mutation |
AAG,AGG |
K552R |
NP_001191065.1 |
NM_001204137.1 |
1885 |
Missense Mutation |
AAG,AGG |
K577R |
NP_001191066.1 |
NM_001204138.1 |
1885 |
Missense Mutation |
AAG,AGG |
K576R |
NP_001191067.1 |
NM_001204139.1 |
1885 |
Missense Mutation |
AAG,AGG |
K552R |
NP_001191068.1 |
NM_001204140.1 |
1885 |
Missense Mutation |
AAG,AGG |
K521R |
NP_001191069.1 |
NM_001204141.1 |
1885 |
Missense Mutation |
AAG,AGG |
K502R |
NP_001191070.1 |
NM_001204142.1 |
1885 |
Missense Mutation |
AAG,AGG |
K506R |
NP_001191071.1 |
NM_001204143.1 |
1885 |
Missense Mutation |
AAG,AGG |
K450R |
NP_001191072.1 |
NM_001204151.2 |
1885 |
Missense Mutation |
AAG,AGG |
K483R |
NP_001191080.1 |
NM_001323942.1 |
1885 |
Missense Mutation |
AAG,AGG |
K577R |
NP_001310871.1 |
NM_001323947.1 |
1885 |
Missense Mutation |
AAG,AGG |
K562R |
NP_001310876.1 |
NM_001323949.1 |
1885 |
Missense Mutation |
AAG,AGG |
K408R |
NP_001310878.1 |
NM_001323950.1 |
1885 |
Missense Mutation |
AAG,AGG |
K536R |
NP_001310879.1 |
NM_001323951.1 |
1885 |
Missense Mutation |
AAG,AGG |
K537R |
NP_001310880.1 |
NM_001323952.1 |
1885 |
Missense Mutation |
AAG,AGG |
K471R |
NP_001310881.1 |
NM_001323953.1 |
1885 |
Missense Mutation |
AAG,AGG |
K314R |
NP_001310882.1 |
NM_001323954.1 |
1885 |
Missense Mutation |
AAG,AGG |
K473R |
NP_001310883.1 |
NM_002384.2 |
1885 |
Missense Mutation |
AAG,AGG |
K450R |
NP_002375.1 |
NM_015844.2 |
1885 |
Missense Mutation |
AAG,AGG |
K496R |
NP_056669.2 |
NM_015845.3 |
1885 |
Missense Mutation |
AAG,AGG |
K483R |
NP_056670.2 |
NM_015846.3 |
1885 |
Missense Mutation |
AAG,AGG |
K552R |
NP_056671.2 |
NM_015847.3 |
1885 |
Missense Mutation |
AAG,AGG |
K503R |
NP_056723.2 |
XM_005258271.2 |
1885 |
Missense Mutation |
AAG,AGG |
K552R |
XP_005258328.1 |
XM_006722456.2 |
1885 |
Missense Mutation |
AAG,AGG |
K577R |
XP_006722519.1 |
XM_011525991.1 |
1885 |
Missense Mutation |
AAG,AGG |
K531R |
XP_011524293.1 |
XM_011525993.2 |
1885 |
Missense Mutation |
AAG,AGG |
K577R |
XP_011524295.1 |
XM_011525994.2 |
1885 |
Missense Mutation |
AAG,AGG |
K577R |
XP_011524296.1 |
XM_011525998.2 |
1885 |
Missense Mutation |
AAG,AGG |
K551R |
XP_011524300.1 |
XM_011525999.1 |
1885 |
Missense Mutation |
AAG,AGG |
K506R |
XP_011524301.1 |
XM_011526001.1 |
1885 |
Missense Mutation |
AAG,AGG |
K496R |
XP_011524303.1 |
XM_011526002.1 |
1885 |
Missense Mutation |
AAG,AGG |
K531R |
XP_011524304.1 |
XM_011526003.2 |
1885 |
Missense Mutation |
AAG,AGG |
K531R |
XP_011524305.1 |
XM_011526006.1 |
1885 |
Missense Mutation |
AAG,AGG |
K450R |
XP_011524308.1 |
XM_011526007.1 |
1885 |
Missense Mutation |
AAG,AGG |
K506R |
XP_011524309.1 |
XM_017025751.1 |
1885 |
Missense Mutation |
AAG,AGG |
K562R |
XP_016881240.1 |
XM_017025752.1 |
1885 |
Missense Mutation |
AAG,AGG |
K537R |
XP_016881241.1 |
XM_017025753.1 |
1885 |
Missense Mutation |
AAG,AGG |
K552R |
XP_016881242.1 |
XM_017025754.1 |
1885 |
Missense Mutation |
AAG,AGG |
K562R |
XP_016881243.1 |
XM_017025755.1 |
1885 |
Missense Mutation |
AAG,AGG |
K552R |
XP_016881244.1 |
XM_017025756.1 |
1885 |
Missense Mutation |
AAG,AGG |
K531R |
XP_016881245.1 |
XM_017025757.1 |
1885 |
Missense Mutation |
AAG,AGG |
K562R |
XP_016881246.1 |
XM_017025758.1 |
1885 |
Missense Mutation |
AAG,AGG |
K562R |
XP_016881247.1 |
XM_017025759.1 |
1885 |
Missense Mutation |
AAG,AGG |
K562R |
XP_016881248.1 |
XM_017025760.1 |
1885 |
Missense Mutation |
AAG,AGG |
K552R |
XP_016881249.1 |
XM_017025761.1 |
1885 |
Missense Mutation |
AAG,AGG |
K531R |
XP_016881250.1 |
XM_017025762.1 |
1885 |
Missense Mutation |
AAG,AGG |
K537R |
XP_016881251.1 |
XM_017025763.1 |
1885 |
Missense Mutation |
AAG,AGG |
K536R |
XP_016881252.1 |
XM_017025764.1 |
1885 |
Missense Mutation |
AAG,AGG |
K531R |
XP_016881253.1 |
XM_017025765.1 |
1885 |
Missense Mutation |
AAG,AGG |
K496R |
XP_016881254.1 |
XM_017025766.1 |
1885 |
Missense Mutation |
AAG,AGG |
K521R |
XP_016881255.1 |
XM_017025767.1 |
1885 |
Missense Mutation |
AAG,AGG |
K521R |
XP_016881256.1 |
XM_017025768.1 |
1885 |
Missense Mutation |
AAG,AGG |
K496R |
XP_016881257.1 |
XM_017025769.1 |
1885 |
Missense Mutation |
AAG,AGG |
K506R |
XP_016881258.1 |
XM_017025770.1 |
1885 |
Missense Mutation |
AAG,AGG |
K496R |
XP_016881259.1 |
XM_017025771.1 |
1885 |
Missense Mutation |
AAG,AGG |
K496R |
XP_016881260.1 |
XM_017025772.1 |
1885 |
Missense Mutation |
AAG,AGG |
K481R |
XP_016881261.1 |
XM_017025773.1 |
1885 |
Missense Mutation |
AAG,AGG |
K473R |
XP_016881262.1 |
XM_017025774.1 |
1885 |
Missense Mutation |
AAG,AGG |
K473R |
XP_016881263.1 |
XM_017025775.1 |
1885 |
Missense Mutation |
AAG,AGG |
K450R |
XP_016881264.1 |
XM_017025776.1 |
1885 |
Missense Mutation |
AAG,AGG |
K450R |
XP_016881265.1 |
XM_017025777.1 |
1885 |
Missense Mutation |
AAG,AGG |
K506R |
XP_016881266.1 |
View Full Product Details