Product Details

SNP ID
rs200801352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54269655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGACGTGGGGGCGGCAGCCAGC[A/T]CGCAGGGTGCGCCGCAGCCAGAGGA
Phenotype
MIM: 605252
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
POLI PubMed Links

Gene Details

Gene
POLI
Gene Name
DNA polymerase iota
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007195.2 98 Missense Mutation ACG,TCG T37S NP_009126.2
XM_005258188.3 98 UTR 5 XP_005258245.1
XM_005258192.4 98 Missense Mutation ACG,TCG T37S XP_005258249.1
XM_006722378.2 98 Missense Mutation ACG,TCG T37S XP_006722441.1
XM_011525796.2 98 UTR 5 XP_011524098.1
XM_011525797.1 98 Missense Mutation ACG,TCG T37S XP_011524099.1

View Full Product Details