Product Details

SNP ID
rs200825989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:31830787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTGATGATGATCAGGGCAGGCA[C/T]GGAATTCTGCTGACTTGTTTCAAAC
Phenotype
MIM: 614136
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC8 PubMed Links

Gene Details

Gene
TRAPPC8
Gene Name
trafficking protein particle complex 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014939.3 4584 Missense Mutation ATG,GTG M1426V NP_055754.2
XM_005258233.2 4584 Missense Mutation ATG,GTG M1427V XP_005258290.1
XM_005258234.2 4584 Missense Mutation ATG,GTG M1397V XP_005258291.1
XM_005258235.3 4584 Missense Mutation ATG,GTG M1152V XP_005258292.1
XM_006722420.3 4584 Intron XP_006722483.1
XM_011525884.2 4584 UTR 3 XP_011524186.1
XM_017025615.1 4584 Missense Mutation ATG,GTG M1401V XP_016881104.1
XM_017025616.1 4584 Missense Mutation ATG,GTG M1396V XP_016881105.1
XM_017025617.1 4584 UTR 3 XP_016881106.1

View Full Product Details