Product Details

SNP ID
rs2230062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33388071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCGCAGGCCGGGCTCGTCGATG[T/C]GCTCCACGCCCTGTGGGGAACAGAG
Phenotype
MIM: 138972 MIM: 613230
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CEBPG PubMed Links

Gene Details

Gene
CEBPG
Gene Name
CCAAT/enhancer binding protein gamma
There are no transcripts associated with this gene.

Gene
PEPD
Gene Name
peptidase D
There are no transcripts associated with this gene.

View Full Product Details