Product Details

SNP ID
rs200956214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:36129945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCAGTTGGCGACATGGTGGCACC[C/G]GTGCTGGAGACTTCTCACGTGTTTT
Phenotype
MIM: 616054 MIM: 608731
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ELP2 PubMed Links

Gene Details

Gene
ELP2
Gene Name
elongator acetyltransferase complex subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242875.2 72 Silent Mutation CCC,CCG P4P NP_001229804.1
NM_001242876.2 72 Silent Mutation CCC,CCG P4P NP_001229805.1
NM_001242877.2 72 Silent Mutation CCC,CCG P4P NP_001229806.1
NM_001242878.2 72 Silent Mutation CCC,CCG P4P NP_001229807.1
NM_001242879.2 72 Silent Mutation CCC,CCG P4P NP_001229808.1
NM_001324465.1 72 Silent Mutation CCC,CCG P4P NP_001311394.1
NM_001324466.1 72 Silent Mutation CCC,CCG P4P NP_001311395.1
NM_001324467.1 72 Silent Mutation CCC,CCG P4P NP_001311396.1
NM_001324468.1 72 UTR 5 NP_001311397.1
NM_018255.3 72 Silent Mutation CCC,CCG P4P NP_060725.1
Gene
SLC39A6
Gene Name
solute carrier family 39 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099406.1 72 Intron NP_001092876.1
NM_012319.3 72 Intron NP_036451.3
XM_011525900.1 72 Intron XP_011524202.1
XM_011525901.1 72 Intron XP_011524203.1

View Full Product Details