Product Details

SNP ID
rs201414884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50227481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTCCTGGACCTTGTCCAAACAC[A/G]TCTTGTTTGCTGCTACACCTGCTTC
Phenotype
MIM: 614759
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP53 PubMed Links

Gene Details

Gene
CFAP53
Gene Name
cilia and flagella associated protein 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145020.4 1563 Missense Mutation ACG,ATG T482M NP_659457.2

View Full Product Details