Product Details

SNP ID
rs201654885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:80160143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGCCGCCGCGCACCACGTTGTT[G/T]CGCTGGTTGGCGGGCTTGACGGTGA
Phenotype
MIM: 608976
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PARD6G PubMed Links

Gene Details

Gene
PARD6G
Gene Name
par-6 family cell polarity regulator gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032510.3 925 Silent Mutation CGA,CGC R253R NP_115899.1
Gene
PARD6G-AS1
Gene Name
PARD6G antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details