Product Details

SNP ID
rs201659207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:47033111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGGCCGCGGGCGCCGCGTGCTC[A/C]TTGCTGCTGCTCAGGCAGGGGTTGG
Phenotype
MIM: 614697 MIM: 609522
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KATNAL2 PubMed Links

Gene Details

Gene
KATNAL2
Gene Name
katanin catalytic subunit A1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031303.2 2507 Intron NP_112593.2
XM_005258357.4 2507 Intron XP_005258414.1
XM_005258358.4 2507 Intron XP_005258415.1
XM_005258361.3 2507 Intron XP_005258418.1
XM_006722554.3 2507 Intron XP_006722617.1
XM_011526219.2 2507 Intron XP_011524521.1
XM_011526220.1 2507 Intron XP_011524522.1
XM_011526221.2 2507 Intron XP_011524523.1
XM_011526223.2 2507 Intron XP_011524525.1
XM_017026028.1 2507 Intron XP_016881517.1
XM_017026029.1 2507 Intron XP_016881518.1
XM_017026030.1 2507 Intron XP_016881519.1
XM_017026031.1 2507 Intron XP_016881520.1
XM_017026032.1 2507 Intron XP_016881521.1
Gene
TCEB3B
Gene Name
transcription elongation factor B subunit 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016427.2 2507 Missense Mutation NP_057511.2
Gene
TCEB3C
Gene Name
transcription elongation factor B subunit 3C
There are no transcripts associated with this gene.

Gene
TCEB3CL
Gene Name
transcription elongation factor B subunit 3C like
There are no transcripts associated with this gene.

View Full Product Details