Product Details

SNP ID
rs201480007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:49490478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAAGTTCAGCATTACTCTCTGC[A/G]TTTTTAAGCATGTGCAGCAAAAATT
Phenotype
MIM: 603661
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C18orf32 PubMed Links

Gene Details

Gene
C18orf32
Gene Name
chromosome 18 open reading frame 32
There are no transcripts associated with this gene.

Gene
LOC101928144
Gene Name
uncharacterized LOC101928144
There are no transcripts associated with this gene.

Gene
MIR1539
Gene Name
microRNA 1539
There are no transcripts associated with this gene.

Gene
RPL17
Gene Name
ribosomal protein L17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000985.4 304 Silent Mutation AAC,AAT N97N NP_000976.1
NM_001035006.2 304 Intron NP_001030178.1
NM_001199340.1 304 Intron NP_001186269.1
NM_001199341.1 304 Intron NP_001186270.1
NM_001199342.1 304 Silent Mutation AAC,AAT N97N NP_001186271.1
NM_001199343.1 304 Silent Mutation AAC,AAT N97N NP_001186272.1
NM_001199344.1 304 Intron NP_001186273.1
NM_001199345.1 304 Intron NP_001186274.1
Gene
RPL17-C18orf32
Gene Name
RPL17-C18orf32 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199355.1 304 Silent Mutation AAC,AAT N97N NP_001186284.1
NM_001199356.1 304 Silent Mutation AAC,AAT N59N NP_001186285.1
Gene
SNORD58A
Gene Name
small nucleolar RNA, C/D box 58A
There are no transcripts associated with this gene.

Gene
SNORD58B
Gene Name
small nucleolar RNA, C/D box 58B
There are no transcripts associated with this gene.

Gene
SNORD58C
Gene Name
small nucleolar RNA, C/D box 58C
There are no transcripts associated with this gene.

View Full Product Details