Product Details

SNP ID
rs201912542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:47157454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTACCTCTCATCACGGTTCTTA[C/G]AGATCGAATAAGGTTCATTAGCTGT
Phenotype
MIM: 609382
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HDHD2 PubMed Links

Gene Details

Gene
HDHD2
Gene Name
haloacid dehalogenase like hydrolase domain containing 2
There are no transcripts associated with this gene.

Gene
IER3IP1
Gene Name
immediate early response 3 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016097.4 272 Missense Mutation CTA,GTA L59V NP_057181.1

View Full Product Details