Product Details

SNP ID
rs201398146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9887906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGGCTGGTGGCTGTGGACTTCT[C/T]GGCCACGTGGTGTGGGCCCTGCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXNDC2 PubMed Links

Gene Details

Gene
TXNDC2
Gene Name
thioredoxin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098529.1 1876 Missense Mutation TCG,TTG S476L NP_001091999.1
NM_032243.5 1876 Missense Mutation TCG,TTG S409L NP_115619.4
XM_017026041.1 1876 Missense Mutation TCG,TTG S409L XP_016881530.1

View Full Product Details