Product Details

SNP ID
rs201172534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9886622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTAGTCCTGTCCAGCAACGTGC[C/T]TCTCCTGGCCCTAGAGTTCTTGGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXNDC2 PubMed Links

Gene Details

Gene
TXNDC2
Gene Name
thioredoxin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098529.1 592 Missense Mutation CCT,CTT P48L NP_001091999.1
NM_032243.5 592 UTR 5 NP_115619.4
XM_017026041.1 592 UTR 5 XP_016881530.1

View Full Product Details