Product Details

SNP ID
rs199847708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9857282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATACCCAGCTTCTGGGCGCAGGC[A/G]TGGAGCCGGGCCTCCAGGGCCATCA
Phenotype
MIM: 601052
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLFM2 PubMed Links

Gene Details

Gene
OLFM2
Gene Name
olfactomedin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304347.1 721 Silent Mutation CAC,CAT H211H NP_001291276.1
NM_001304348.1 721 Silent Mutation CAC,CAT H109H NP_001291277.1
NM_058164.3 721 Silent Mutation CAC,CAT H187H NP_477512.1
Gene
PIN1
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 1
There are no transcripts associated with this gene.

View Full Product Details