Product Details

SNP ID
rs200158223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38945034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGGCTGAGAACTTGACCACTT[C/T]CTTTTCATACACATCCAGAAGGCGG
Phenotype
MIM: 609094
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO17 PubMed Links

Gene Details

Gene
FBXO17
Gene Name
F-box protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024907.6 760 Missense Mutation AAA,GAA K210E NP_079183.4
NM_148169.2 760 Missense Mutation AAA,GAA K219E NP_680474.1

View Full Product Details