Product Details

SNP ID
rs200352688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3959301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCGCCTGCCGCTTGAGCGCCT[C/T]GGTCTTGAGCAGCTCCTGCCGTAGC
Phenotype
MIM: 603289
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAPK3 PubMed Links

Gene Details

Gene
DAPK3
Gene Name
death associated protein kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001348.2 1462 Missense Mutation AAG,GAG K389E NP_001339.1
XM_005259508.1 1462 Missense Mutation AAG,GAG K389E XP_005259565.1
Gene
MIR637
Gene Name
microRNA 637
There are no transcripts associated with this gene.

View Full Product Details