Product Details

SNP ID
rs199674324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39837823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGCCAGAGCGGTGGGAGAACTC[A/G]ACTGCATAGACTAGACCATCCTAAA
Phenotype
MIM: 604556 MIM: 134795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYRK1B PubMed Links

Gene Details

Gene
DYRK1B
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1B
There are no transcripts associated with this gene.

Gene
FBL
Gene Name
fibrillarin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001436.3 465 Silent Mutation GTC,GTT V190V NP_001427.2
XM_005258651.2 465 Silent Mutation GTC,GTT V189V XP_005258708.1
XM_011526623.2 465 Silent Mutation GTC,GTT V133V XP_011524925.1
Gene
MIR6719
Gene Name
microRNA 6719
There are no transcripts associated with this gene.

View Full Product Details